Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tubgcp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146190.2 | NP_666302.2 | 12 | 1524 | ||
GenBank mRNA | AK032031.1 | - | 12 | 1484 | ||
AK044444.1 | - | 12 | 1410 | |||
AK044887.1 | - | 12 | 1508 | |||
AK051326.1 | - | 12 | 1519 | |||
AK086158.1 | - | 12 | 1485 | |||
AK134963.1 | - | 4 | 642 | |||
AK151454.1 | - | 12 | 1482 | |||
AK152033.1 | - | 12 | 1482 | |||
AK153331.1 | - | 12 | 1482 | |||
AK153357.1 | - | 12 | 1482 | |||
AK160939.1 | - | 12 | 1438 | |||
AK161681.1 | - | 12 | 1482 | |||
AK169443.1 | - | 3 | 868 | |||
AK173291.1 | - | 9 | 901 | |||
BC027075.1 | - | 4 | 587 | 5355335 |