Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abca9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147220.2 | NP_671753.2 | 4 | 609 | ||
GenBank mRNA | AF491299.1 | - | 4 | 609 | ||
AK085555.1 | - | 4 | 513 |