Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dner | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152915.1 | NP_690879.1 | 13 | 2892 | ||
GenBank mRNA | AB067650.1 | - | 13 | 2541 | ||
AF370126.1 | - | 13 | 2548 | |||
AK044597.1 | - | 13 | 2655 | |||
AK152755.1 | - | 8 | 1294 | |||
AK153235.1 | - | 8 | 1294 | |||
AY032924.1 | - | 13 | 2548 | |||
BC022636.1 | - | 8 | 1355 | |||
BC034634.1 | - | 13 | 2892 | 5365190 |