Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152944.1 | NP_694423.1 | 2 | 270 | ||
GenBank mRNA | AF507967.1 | - | 2 | 255 | ||
AY124569.1 | - | 2 | 270 | |||
BC106860.2 | - | 2 | 270 | 40047272 | ||
BC106861.2 | - | 2 | 270 | 40047279 | ||
BC111522.1 | - | 1 | 1077 | 40047286 |