Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ebf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110513.1 | NP_001103983.1 | 9 | 900 | ||
GenBank mRNA | AF387630.1 | - | 9 | 900 | ||
AF387631.1 | - | 9 | 900 | |||
AF387632.1 | - | 9 | 900 | |||
AF387633.1 | - | 9 | 900 | |||
AF387634.1 | - | 9 | 900 | |||
BC138119.1 | - | 9 | 900 | 8861138 | ||
BC145155.1 | - | 10 | 986 | 9053706 |