Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapre2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162941.1 | NP_001156413.1 | 4 | 420 | ||
NM_001162942.1 | NP_001156414.1 | 4 | 363 | |||
NM_153058.4 | NP_694698.3 | 3 | 471 | |||
GenBank mRNA | AI325879.1 | - | 4 | 357 | ||
AK028823.1 | - | 4 | 421 | |||
AK038562.1 | - | 3 | 458 | |||
AK040767.1 | - | 3 | 446 | |||
AK045493.1 | - | 3 | 354 | |||
AK045904.1 | - | 3 | 446 | |||
AK046095.1 | - | 3 | 413 | |||
AK046221.1 | - | 3 | 438 | |||
AK148954.1 | - | 3 | 446 | |||
AK168834.1 | - | 5 | 305 | |||
AK188282.1 | - | 160 | ||||
AK192344.1 | - | 1 | 55 | |||
BC025804.1 | - | 3 | 418 | 5101879 | ||
BC027056.1 | - | 3 | 437 | 5361238 | ||
BC028987.1 | - | 3 | 393 | 5362960 | ||
BC035254.1 | - | 3 | 423 | 5363176 |