Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ak5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081277.1 | NP_001074746.1 | 13, 14 | 1900 | ||
GenBank mRNA | AB060081.1 | - | 4, 5 | 512 | ||
AK053807.1 | - | 13, 14 | 1898 |