Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153082.4 | NP_694722.2 | 3, 4 | 461 | ||
GenBank mRNA | AK049300.1 | - | 3, 4 | 407 | ||
AY035893.1 | - | 3, 4 | 248 | |||
BC042643.1 | - | 3, 4 | 376 | 5357595 | ||
BC051140.1 | - | 3, 4 | 423 | 6595101 | ||
CB247441.1 | - | 2, 3 | 175 |