Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctdsp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153088.2 | NP_694728.1 | 7 | 1511 | ||
GenBank mRNA | AK089478.1 | - | 7 | 1419 | ||
AK167390.1 | - | 7 | 1269 | |||
AY028804.1 | - | 7 | 1536 | |||
BC049184.1 | - | 7 | 1371 | 6439402 | ||
BC058085.1 | - | 6 | 1698 | |||
BC065158.1 | - | 7 | 1246 | 5717102 | ||
BC079638.1 | - | 7 | 1246 | 5717102 |