Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AF366264 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153093.3 | NP_694733.3 | 1 | 1248 | ||
GenBank mRNA | AK030169.1 | - | 1 | 814 |
Gene Symbol: 4930444G20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053264.2 | NP_444494.1 | 1 | 1227 | ||
GenBank mRNA | AK015380.1 | - | 1 | 814 |
Gene Symbol: Gm5415 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164286.1 | NP_001157758.1 | 1 | 1642 | ||
GenBank mRNA | AK029494.1 | - | 1 | 1644 |
Gene Symbol: Gm9839 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199956.1 | NP_001186885.1 | 1 | 1176 |