Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wipf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289722.1 | NP_001276651.1 | 4 | 519 | ||
NM_001289723.1 | NP_001276652.1 | 4 | 484 | |||
NM_153138.4 | NP_694778.1 | 4 | 514 | |||
GenBank mRNA | AJ437262.1 | - | 3 | 315 | ||
AK031046.1 | - | 4 | 481 | |||
AK156547.1 | - | 4 | 519 | |||
BC049788.1 | - | 4 | 437 | 6514406 |