Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc36a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153139.4 | NP_694779.3 | 9 | 1105 | ||
GenBank mRNA | AF453743.1 | - | 8 | 1091 | ||
AY211262.1 | - | 9 | 1075 | |||
BC138556.1 | - | 9 | 1069 | 8861576 | ||
BC138558.1 | - | 9 | 1069 | 8861578 |