Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ggnbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153144.2 | NP_694784.2 | 3, 4 | 386 | ||
GenBank mRNA | AB064543.1 | - | 3, 4 | 386 | ||
AY633741.1 | - | 3, 4 | 386 | |||
BC060738.1 | - | 2, 3 | 342 | |||
BC139772.1 | - | 2, 3 | 292 | 40111779 | ||
BC145605.1 | - | 2, 3 | 294 | 9054171 | ||
BC150801.1 | - | 2, 3 | 294 | 9087712 |