Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abca8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153145.4 | NP_694785.3 | 3 | 201 | ||
GenBank mRNA | AB255881.1 | - | 2 | 120 | ||
AB255882.1 | - | 3 | 208 | |||
AF498360.1 | - | 2 | 126 | |||
AY732492.1 | - | 2 | 126 |