Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278784.1 | NP_001265713.1 | 7 | 805 | ||
NM_005827.2 | NP_005818.2 | 7 | 830 | |||
XM_005256929.3 | XP_005256986.1 | 8 | 745 | |||
XM_006721632.3 | XP_006721695.1 | 8 | 949 | |||
XM_011524179.2 | XP_011522481.1 | 6 | 715 | |||
XM_011524180.2 | XP_011522482.1 | 7 | 681 | |||
GenBank mRNA | AK098056.1 | - | 5 | 1905 | ||
AK124975.1 | - | 1 | 751 | |||
AK293563.1 | - | 7 | 830 | |||
BC011888.2 | - | 7 | 809 | 3832062 | ||
BM476321.1 | - | 3 | 240 | |||
BX641166.1 | - | 7 | 1084 | |||
CR745911.1 | - | 5 | 414 | |||
CX164572.1 | - | 5 | 437 | |||
D87989.1 | - | 7 | 799 |