Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txnrd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178058.1 | NP_001171529.1 | 3 | 536 | ||
NM_001178059.1 | NP_001171530.1 | 3 | 536 | |||
NM_001178060.1 | NP_001171531.1 | 3 | 536 | |||
NM_153162.3 | NP_694802.2 | 3 | 536 | |||
GenBank mRNA | AF349659.1 | - | 3 | 445 | ||
AK012699.1 | - | 3 | 529 | |||
AK041046.1 | - | 3 | 537 | |||
AK080362.1 | - | 3 | 527 | |||
AK134987.1 | - | 3 | 527 | |||
AK213765.1 | - | 2 | 124 | |||
BC076605.1 | - | 3 | 494 | 30628949 |