Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153391.2 | NP_700440.2 | 6 | 572 | ||
GenBank mRNA | AK043836.1 | - | 6 | 529 | ||
AK052303.1 | - | 6 | 552 | |||
AK147970.1 | - | 6 | 572 | |||
AK166172.1 | - | 6 | 538 | |||
AY029258.1 | - | 6 | 642 | |||
BC060134.1 | - | 6 | 552 | |||
BC082301.1 | - | 5 | 452 |