Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153391.2 | NP_700440.2 | 17 | 1894 | ||
GenBank mRNA | AK052303.1 | - | 17 | 1874 | ||
AK129413.1 | - | 9 | 983 | |||
AK147970.1 | - | 17 | 1894 | |||
AK166172.1 | - | 17 | 1860 | |||
AY029258.1 | - | 17 | 1964 | |||
BC046432.1 | - | 2 | 96 | |||
BC060134.1 | - | 17 | 1888 | |||
BC082301.1 | - | 16 | 1774 |