Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012392.1 | NP_001012392.1 | 5, 6 | 639 | ||
NM_153418.2 | NP_700467.2 | 5, 6 | 602 | |||
GenBank mRNA | AK146605.1 | - | 5, 6 | 635 | ||
BC060982.1 | - | 5, 6 | 639 | 30300112 | ||
BC089529.1 | - | 639 | ||||
BC100436.1 | - | 5, 6 | 634 | 30298932 | ||
U46068.3 | - | 5, 6 | 603 |