Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Insig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153526.5 | NP_705746.1 | 5 | 1562 | ||
GenBank mRNA | AK088380.1 | - | 5 | 1609 | ||
AK135941.1 | - | 5 | 1613 | |||
AK135989.1 | - | 5 | 1615 | |||
AK139913.1 | - | 5 | 1612 | |||
AK151186.1 | - | 5 | 1612 | |||
BC079559.1 | - | 5 | 1562 | |||
BC132167.1 | - | 5 | 1570 | 40130444 | ||
BC132499.1 | - | 5 | 1570 | 40130776 |