Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brinp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145807.1 | NP_001139279.1 | 4 | 609 | ||
NM_153539.3 | NP_705767.3 | 4 | 967 | |||
GenBank mRNA | AK046878.1 | - | 4 | 968 | ||
AK083233.1 | - | 5 | 658 | |||
BC030498.1 | - | 3 | 585 | 5400263 | ||
BC099956.1 | - | 4 | 944 | 30101722 |