Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153545.2 | NP_705773.2 | 18 | 2011 | ||
GenBank mRNA | AK081566.1 | - | 17 | 1939 | ||
AK153361.1 | - | 18 | 2012 | |||
BC023296.1 | - | 18 | 1991 | 3662961 | ||
CN539620.1 | - | 3 | 261 |