Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dirc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153550.3 | NP_705778.1 | 9 | 1586 | ||
GenBank mRNA | AK034214.1 | - | 9 | 1586 | ||
AK172331.1 | - | 9 | 1601 | |||
BC037460.1 | - | 9 | 1560 | 4190539 |