Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gbp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153564.2 | NP_705792.2 | 11 | 2419 | ||
GenBank mRNA | AF422243.1 | - | 11 | 2401 | ||
AF487898.1 | - | 10 | 3015 | |||
AK030414.1 | - | 12 | 3359 | |||
BC058555.1 | - | 11 | 2453 | 5293575 |