Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153573.1 | NP_705801.1 | 2 | 393 | ||
GenBank mRNA | AK030620.1 | - | 2 | 443 | ||
AK081692.1 | - | 2 | 420 | |||
AK163384.1 | - | 539 | ||||
BC024846.1 | - | 730 | ||||
BC029109.1 | - | 2 | 393 | 3985764 | ||
BC034313.1 | - | 2 | 418 | 5365046 |