Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syne4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290565.1 | NP_001277494.1 | 7 | 1075 | ||
NM_153577.2 | NP_705805.1 | 8 | 1324 | |||
GenBank mRNA | AK002612.1 | - | 1697 | |||
BC023803.1 | - | 8 | 1311 | 5352953 |