Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdkl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166653.1 | NP_001160125.1 | 7 | 1072 | ||
NM_001166654.1 | NP_001160126.1 | 7 | 1162 | |||
NM_001166655.1 | NP_001160127.1 | 7 | 1165 | |||
NM_001166656.1 | NP_001160128.1 | 7 | 1072 | |||
NM_001166657.1 | NP_001160129.1 | 7 | 1162 | |||
NM_153785.4 | NP_722480.3 | 7 | 1162 | |||
GenBank mRNA | AK045036.1 | - | 6 | 1521 | ||
AK045356.1 | - | 7 | 1062 | |||
AK046394.1 | - | 7 | 1163 | |||
AK050990.1 | - | 7 | 1162 | |||
AK078804.1 | - | 7 | 1054 | |||
AK090045.1 | - | 7 | 948 | |||
AK166428.1 | - | 7 | 1004 | |||
BC028871.1 | - | 7 | 1034 | 4241862 |