Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153788.3 | NP_722483.2 | 4, 5 | 497 | ||
GenBank mRNA | AK133473.1 | - | 3, 4 | 721 | ||
AK138736.1 | - | 4, 5 | 489 | |||
AK154210.1 | - | 4, 5 | 462 | |||
AK154449.1 | - | 4, 5 | 500 | |||
AK155001.1 | - | 4, 5 | 518 | |||
BC031462.1 | - | 4, 5 | 497 | 4017986 |