Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam134a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170755.2 | NP_739561.2 | 9 | 1210 | ||
GenBank mRNA | AK034667.1 | - | 9 | 1213 | ||
AK152268.1 | - | 9 | 925 | |||
AK158940.1 | - | 9 | 979 | |||
AK160400.1 | - | 9 | 1164 | |||
AK165957.1 | - | 9 | 1189 | |||
AK172093.1 | - | 9 | 1207 | |||
AK191491.1 | - | 2 | 186 | |||
AK210282.1 | - | 2 | 186 | |||
AK210397.1 | - | 2 | 186 | |||
AK218481.1 | - | 2 | 186 | |||
BC026519.1 | - | 5 | 561 | |||
BC038286.1 | - | 5 | 596 | 4221441 | ||
BC070406.1 | - | 9 | 1202 | 5707319 |