Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd300a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170758.3 | NP_739564.1 | 7 | 1637 | ||
GenBank mRNA | AB065156.1 | - | 7 | 1356 | ||
AB091765.1 | - | 7 | 1363 | |||
AB091766.1 | - | 7 | 1359 | |||
AB095675.1 | - | 7 | 1460 | |||
AK045869.1 | - | 7 | 1637 | |||
BC139296.1 | - | 7 | 1573 | 8862318 | ||
BC139297.1 | - | 7 | 1573 | 8862319 |