Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: U2af1l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170760.3 | NP_739566.1 | 8 | 714 | ||
GenBank mRNA | BC051477.1 | - | 7 | 525 | ||
BC060972.1 | - | 8 | 714 | 6593404 |