Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172049.2 | NP_742046.2 | 5 | 1131 | ||
GenBank mRNA | AK133493.1 | - | 5 | 1131 | ||
AK160817.1 | - | 5 | 1123 | |||
AK166323.1 | - | 5 | 1148 | |||
AK170496.1 | - | 5 | 1114 | |||
BC024093.1 | - | 5 | 1103 | 5116657 |