Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172133.1 | NP_742145.1 | 2 | 238 | ||
GenBank mRNA | AK050079.1 | - | 2 | 296 | ||
AK050398.1 | - | 2 | 287 | |||
AK053152.1 | - | 2 | 282 | |||
AK139005.1 | - | 2 | 290 | |||
AK140864.1 | - | 2 | 269 | |||
BC027165.1 | - | 2 | 238 | 4951680 |