Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172133.1 | NP_742145.1 | 6 | 688 | ||
GenBank mRNA | AK050079.1 | - | 6 | 746 | ||
AK050398.1 | - | 6 | 737 | |||
AK053152.1 | - | 6 | 732 | |||
AK139005.1 | - | 6 | 740 | |||
AK140864.1 | - | 6 | 719 | |||
BC027165.1 | - | 6 | 688 | 4951680 |