Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eva1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172145.3 | NP_742157.1 | 3 | 1143 | ||
GenBank mRNA | AK076006.1 | - | 3 | 1145 | ||
AK077984.1 | - | 3 | 1473 | |||
BC029204.1 | - | 2 | 776 | 3964022 |