Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dync2li1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172256.1 | NP_758460.1 | 4, 5 | 320 | ||
GenBank mRNA | AK008822.1 | - | 3, 4 | 269 | ||
AK160259.1 | - | 4, 5 | 334 | |||
BC030450.1 | - | 4, 5 | 322 | 5400492 | ||
BC039070.1 | - | 4, 5 | 320 | 3482503 |