Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sidt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289668.1 | NP_001276597.1 | 27 | 4154 | ||
NM_172257.4 | NP_758461.1 | 26 | 4091 | |||
GenBank mRNA | AK164745.1 | - | 1 | 915 | ||
BC006873.1 | - | 13 | 2428 | |||
BC023957.1 | - | 26 | 4095 | 5321190 | ||
BC051101.1 | - | 27 | 3882 | 5052849 |