Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172269.3 | NP_758473.3 | 4 | 1508 | ||
GenBank mRNA | AK029109.1 | - | 4 | 1485 | ||
AK033333.1 | - | 4 | 1493 | |||
AK036915.1 | - | 4 | 1493 | |||
AK039848.1 | - | 4 | 1493 | |||
AK042471.1 | - | 4 | 1496 | |||
AK044821.1 | - | 4 | 1511 | |||
AK148077.1 | - | 4 | 1500 | |||
AK151113.1 | - | 1 | 130 | |||
AK152700.1 | - | 1 | 130 | |||
AK160689.1 | - | 4 | 1486 | |||
AK173179.1 | - | 1 | 654 | |||
BC026870.1 | - | 1 | 434 | |||
BC036129.1 | - | 4 | 1436 | 4972693 | ||
BC039176.1 | - | 4 | 1415 | 4221232 |