Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172269.3 | NP_758473.3 | 5 | 3789 | ||
GenBank mRNA | AK029109.1 | - | 5 | 3766 | ||
AK033333.1 | - | 5 | 3774 | |||
AK036915.1 | - | 5 | 3774 | |||
AK036947.1 | - | 2 | 1951 | |||
AK042471.1 | - | 5 | 3777 | |||
AK148077.1 | - | 5 | 3781 | |||
AK160689.1 | - | 5 | 3767 | |||
AK173179.1 | - | 2 | 2935 | |||
BC026870.1 | - | 2 | 2715 | |||
BC036129.1 | - | 5 | 3717 | 4972693 | ||
BC039176.1 | - | 5 | 3696 | 4221232 |