Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083887.2 | NP_001077356.1 | 2 | 1233 | ||
NM_001285783.1 | NP_001272712.1 | 2 | 1250 | |||
NM_001285784.1 | NP_001272713.1 | 2 | 401 | |||
NM_172272.2 | NP_758476.2 | 2 | 384 | |||
GenBank mRNA | AK154150.1 | - | 2 | 1205 | ||
BC027073.1 | - | 1 | 74 | |||
BC038817.1 | - | 2 | 357 | 3256949 |