Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cc2d2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172274.2 | NP_758478.1 | 13 | 1765 | ||
GenBank mRNA | AK037073.1 | - | 2 | 205 | ||
AK077658.1 | - | 10 | 1240 | |||
BC032290.1 | - | 13 | 1709 | 5369681 |