Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172338.2 | NP_758841.1 | 4 | 338 | ||
GenBank mRNA | AK028692.1 | - | 4 | 343 | ||
AK051048.1 | - | 4 | 339 | |||
AK122407.1 | - | 585 | ||||
AK154946.1 | - | 359 | ||||
AK164023.1 | - | 4 | 354 |