Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Poglut1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300827.1 | NP_001287756.1 | 2 | 411 | ||
NM_172380.4 | NP_759012.1 | 2 | 208 | |||
GenBank mRNA | AK031608.1 | - | 2 | 147 | ||
AK035948.1 | - | 2 | 156 | |||
AK036224.1 | - | 2 | 138 | |||
AK041321.1 | - | 2 | 146 | |||
AK045278.1 | - | 2 | 356 | |||
AK079886.1 | - | 2 | 177 | |||
AK082556.1 | - | 2 | 378 | |||
AK167641.1 | - | 2 | 378 | |||
AK197458.1 | - | 1 | 248 | |||
AK215456.1 | - | 2 | 50 | |||
AK216766.1 | - | 2 | 50 | |||
BC026809.1 | - | 2 | 159 | 4205469 | ||
CB249259.1 | - | 2 | 209 |