Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Poglut1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300827.1 | NP_001287756.1 | 6 | 919 | ||
NM_172380.4 | NP_759012.1 | 6 | 716 | |||
GenBank mRNA | AK031608.1 | - | 6 | 655 | ||
AK035948.1 | - | 6 | 664 | |||
AK036224.1 | - | 6 | 646 | |||
AK041321.1 | - | 6 | 654 | |||
AK045278.1 | - | 6 | 864 | |||
AK079886.1 | - | 6 | 685 | |||
AK167641.1 | - | 6 | 886 | |||
AK204867.1 | - | 4 | 345 | |||
BC026809.1 | - | 6 | 667 | 4205469 |