Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3bgrl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172507.5 | NP_766095.1 | 4 | 1219 | ||
GenBank mRNA | AK049293.1 | - | 1 | 253 | ||
AK076202.1 | - | 4 | 1208 | |||
AK160357.1 | - | 4 | 1208 | |||
BC038473.1 | - | 4 | 1258 | 5008777 |