Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172509.3 | NP_766097.1 | 4 | 1087 | ||
GenBank mRNA | AK029942.1 | - | 4 | 1063 | ||
AK034583.1 | - | 4 | 1063 | |||
AK083583.1 | - | 4 | 1088 | |||
AK140606.1 | - | 4 | 1062 | |||
AK162568.1 | - | 4 | 1116 | |||
BC051544.1 | - | 3 | 630 | |||
BC139070.1 | - | 4 | 989 | 8862092 | ||
BC141201.1 | - | 4 | 989 | 9055813 |