Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbbp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172517.2 | NP_766105.2 | 3 | 236 | ||
GenBank mRNA | AK049247.1 | - | 3 | 223 | ||
AK154782.1 | - | 3 | 237 | |||
AK170520.1 | - | 3 | 235 | |||
AK171177.1 | - | 3 | 241 | |||
AK195903.1 | - | 3 | 112 |