Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172518.3 | NP_766106.2 | 4 | 739 | ||
GenBank mRNA | AK028867.1 | - | 4 | 585 | ||
AK150804.1 | - | 4 | 730 | |||
AK151503.1 | - | 4 | 696 | |||
AK165284.1 | - | 4 | 713 | |||
AK170202.1 | - | 4 | 741 | |||
AK171215.1 | - | 4 | 742 | |||
AK208927.1 | - | 2 | 132 | |||
BC058667.1 | - | 4 | 730 | 6410012 |