Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172518.3 | NP_766106.2 | 4 | 758 | ||
GenBank mRNA | AK028867.1 | - | 4 | 604 | ||
AK150804.1 | - | 4 | 749 | |||
AK151503.1 | - | 4 | 715 | |||
AK165284.1 | - | 4 | 732 | |||
AK170202.1 | - | 4 | 760 | |||
AK171215.1 | - | 4 | 761 | |||
BC058667.1 | - | 4 | 749 | 6410012 |