Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Megf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134399.1 | NP_001127871.1 | 4 | 420 | ||
NM_172522.4 | NP_766110.3 | 4 | 420 | |||
GenBank mRNA | AK032661.1 | - | 3 | 139 | ||
AK048840.1 | - | 4 | 364 | |||
AK051642.1 | - | 4 | 357 | |||
AK122555.2 | - | 4 | 360 | |||
BB650216.1 | - | 4 | 361 | |||
BC151157.1 | - | 4 | 321 | 9088045 | ||
BC172024.1 | - | 4 | 321 | 9054310 | ||
CJ049881.1 | - | 4 | 396 |